Stiripentol 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
471450010
471450050
471450010.EA
81.9
EUR
471450010.
471450050.
Stiripentol 95%
Stiripentol
Formel:
C₁₄H₁₈O₃ Siedepunkt: 365°C (760mmHg) Flammpunkt: 175°C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00869310 CAS-Nummer: 49763-96-4 EINECS: 256-480-9 |
Spezifikation Testergebnisse
Appearance | white to beige crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Melting point | 75°C to 79°C |
Water | ≤5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...