Vardenafil hydrochloride trihydrate 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Vardenafil HCl trihydrate, BAY 38-9456, 2-(2-Ethoxy-5-((4-ethylpiperazin-1-yl)sulphonyl)phenyl)-5-methyl-7-propylimidazo[5,1-f][1,2,4]triazin-4(1H)-one hydrochloride trihydrate

471490010 471492500
471490010.EA 581 EUR
471490010. 471492500.
Vardenafil hydrochloride trihydrate 97%
Vardenafil hydrochloride trihydrate
Formel: C₂₃H₃₉ClN₆O₇S
Molecular Weight: 579.10 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD09027942
CAS-Nummer: 330808-88-3

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white powder
Infrared spectrum Conforms
HPLC ≥96 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR