Prunetin 95%
Lieferant: Thermo Fisher Scientific
471630010
471632500
471630010.EA
222
EUR
471630010.
471632500.
Prunetin 95%
Prunetin
Formel:
C₁₆H₁₂O₅ Siedepunkt: 546°C (760mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016951 CAS-Nummer: 552-59-0 EINECS: 209-018-5 |
Spezifikation Testergebnisse
Appearance | white to light beige crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...