Diisopropyladipate 99%

Lieferant: Thermo Scientific

Synonyme: Adipic acid diisopropyl ester

471870250 471871000
471870250.EA 18.8 EUR
471870250. 471871000.
Diisopropyladipate 99%
Diisopropyl adipate
Formel: C₁₂H₂₂O₄
Molecular Weight: 230,30 g/mol
Siedepunkt: 120°C (760mmHg)
Flammpunkt: 124°C
Lagertemperatur: Raumtemperatur
MDL: MFCD00026391
CAS-Nummer: 6938-94-9
EINECS: 230-072-0

Bestellen

Spezifikation Testergebnisse

Appearance clear colorless to almost colorless liquid
Infrared spectrum Conforms
GC ≥98.5 %
Refractive index 1.4220 to 1.4260 (20°C, 589 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR