Racecadotril 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Benzyl N-[3-(Acetylthio)-2-benzylpropionyl]glycinate, Acetorphan, N-[3-(Acetylthio)-2-benzylpropionyl]glycine Benzyl Ester

471980010 471980050
471980010.EA 49.6 EUR
471980010. 471980050.
Racecadotril 98%
Racecadotril
Formel: C₂₁H₂₃NO₄S
Molecular Weight: 385,48 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00865520
CAS-Nummer: 81110-73-8
UN: 3077
ADR: 9,III
Merck Index: 13,08182

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Loss on drying ≤0.5 % (1 g, 60°C, 4 hr) (vacuum)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR