Cymoxanil 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Cyan-N-[(ethylamino)carbonyl]-2-(methoxyimino)acetamide, 1-(2-Cyano-2-methoxyiminoacetyl)-3-ethylurea, 2-Cyano-N-(ethylcarbamoyl)-2-methoxyiminoacetamide

472070050 472070250
472070050.EA 31 EUR
472070050. 472070250.
Cymoxanil 98%
Cymoxanil
Formel: C₇H₁₀N₄O₃
Molecular Weight: 198,18 g/mol
Flammpunkt: 100°C
Lagertemperatur: Raumtemperatur
MDL: MFCD00137381
CAS-Nummer: 57966-95-7
EINECS: 261-043-0
UN: 3077
ADR: 9,III
Merck Index: 13,02794

Bestellen

Spezifikation Testergebnisse

Appearance white to yellow to orange to brown powder or crystals
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 156°C to 164°C
Water ≤1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR