Regorafenib 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-(4-(3-(4-Chloro-3-(trifluoromethyl)phenyl)ureido)-3-fluorophenoxy)-N-methylpicolinamide, BAY-73-4506
472550010
472551000
472550010.EA
588
EUR
472550010.
472551000.
Regorafenib 97%
Regorafenib
Formel:
C₂₁H₁₅ClF₄N₄O₃ Molecular Weight: 482,8 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD16038047 CAS-Nummer: 755037-03-7 UN: 3077 ADR: 9,III |
Spezifikation Testergebnisse
Appearance | white to almost white solid |
Infrared spectrum | Conforms |
HPLC | ≥96 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...