1H-[1,2,3]Triazole-4-carboxylic acid 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1H-[1,2,3]Triazole-4-carboxylic acid, 3H-[1,2,3]Triazole-4-carboxylic acid, 1H-1,2,3-Triazole-4-carboxylic acid, 3H-1-2-3-Triazole-4-carboxylic-acid, 4-Carboxy-1H-1,2,3-triazole
472600010
472600050
472600010.EA
106
EUR
472600010.
472600050.
1H-[1,2,3]Triazole-4-carboxylic acid 95%
1H-[1,2,3]Triazole-4-carboxylic acid
Formel:
C₃H₃N₃O₂ Molecular Weight: 113,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00196046 CAS-Nummer: 16681-70-2 |
Spezifikation Testergebnisse
Appearance | white to almost white powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...