5-Carboxytetramethylrhodamine (5-TAMRA) 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5-TAMRA, 5-Carboxytetramethylrhodamine, 2-(3,6-Bis(dimethylamino)xanthylium-9-yl)-5-carboxybenzoate, 2-[3,6-Bis(dimethylamino)xanthen-10-ium-9-yl]-5-carboxybenzoate, TAMRA, 5-isomer
472621000
472625000
472621000.EA
82.6
EUR
472621000.
472625000.
5-Carboxytetramethylrhodamine (5-TAMRA) 95%
5-Carboxytetramethylrhodamine (5-TAMRA)
Formel:
C₂₅H₂₂N₂O₅ Molecular Weight: 430.45 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD1073638 CAS-Nummer: 91809-66-4 |
Spezifikation Testergebnisse
Appearance | green to brown powder or crystals |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
UV Lambda max | 539 to 545 nm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...