Pheniramine maleate 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: N,N-Dimethyl-3-phenyl-3-(pyridin-2-yl)propan-1-amine maleate

472670010 472670050
472670010.EA 44.6 EUR
472670010. 472670050.
Pheniramine maleate 98%
Pheniramine maleate
Formel: C₂₀H₂₄N₂O₄
Molecular Weight: 356,422 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00079250
CAS-Nummer: 132-20-7
EINECS: 205-051-4

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white powder or crystals
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR