2,3':4',2''-Terthiophene 95%
Lieferant: Thermo Scientific
|
Achtung
|
472800010
472802500
472800010.EA
241
EUR
472800010.
472802500.
2,3':4',2''-Terthiophene 95%
2,3':4',2''-Terthiophene
Formel:
C₁₂H₈S₃ Molecular Weight: 248,39 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01106341 CAS-Nummer: 101306-11-0 |
Spezifikation Testergebnisse
Appearance | white to almost white solid |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...