AmberChrom™ hydrogen form 50WX8 50-100 mesh
Lieferant: Thermo Fisher Scientific
Gefahr
472910025 472911000 472915000
472910025.EA 1254 EUR
472910025. 472911000. 472915000.
AmberChrom™ hydrogen form 50WX8 50-100 mesh
AmberChrom™ hydrogen form
Lagertemperatur: Raumtemperatur
MDL: MFCD00132726
CAS-Nummer: 69011-22-9
Bestellen
Spezifikation Testergebnisse
Appearance beige to green-beige to light brown beads
Total exchange capacity ≥1.7 eq/l (H form)
Water retention capacity 50 to 56 % (H form)
Particle size 50 to 100 mesh