1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
DOTA, 1,4,7,10-Tetraazacyclododecane-N,N',N'',N'''-tetraacetic acid, 2,2',2'',2'''-(1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetrayl)tetraacetic acid
473010010
473012500
473010010.EA
328
EUR
473010010.
473012500.
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid 98%
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid
Formel:
C₁₆H₂₈N₄O₈ Molecular Weight: 404,42 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00068657 CAS-Nummer: 60239-18-1 |
Spezifikation Testergebnisse
Appearance | white to almost white powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...