Ceresin wax, weiß
Lieferant: Thermo Scientific
473450010
473452500
473450010.EA
96.9
EUR
473450010.
473452500.
Ceresin wax, weiß
Ceresin wax
Lagertemperatur:
Raumtemperatur |
MDL:
MFCD00146361 CAS-Nummer: 8001-75-0 EINECS: 232-290-1 Merck Index: 13,01999 |
Spezifikation Testergebnisse
Appearance | white to almost white pellets |
Infrared spectrum | Conforms |
Melting point | 58.0°C to 65.0°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...