Anilinblau, wasserlöslich
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
CI 42755, Dinatriumhydrogenaminomethyl[[4-[(sulfonatophenyl)amino]phenyl][4-[(sulfonatophenyl)imino]cyclohexa-2,5-dien-1-yliden]methyl]benzolsulfonat
474360050
474360250
474360050.EA
39.4
EUR
474360050.
474360250.
Anilinblau, wasserlöslich
Anilinblau
Formel:
C₃₂H₂₅N₃Na₂O₉S₃ Molecular Weight: 737.74 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036130 CAS-Nummer: 28631-66-5 EINECS: 249-113-9 |
Spezifikation Testergebnisse
Appearance | dark blue to dark brown powder or crystals |
Infrared spectrum | Conforms |
UV Lambda max | 590 to 610 nm (in dilute HCl) |
Absorption Ratio P-15/P+15 | 1 to 1.03 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...