IPTG (Isopropyl-β-D-thiogalactopyranosid) ≥99% dioxanfrei
Lieferant: Thermo Scientific
Synonyme: IPTG
B21149.03 B21149.06
B21149.03EA 90.2 EUR
B21149.03 B21149.06
IPTG (Isopropyl-β-D-thiogalactopyranosid) ≥99% dioxanfrei
IPTG (Isopropyl-β-D-thiogalactopyranosid)
Purity: 99% CAS No.: 367-93-1 Molecular Formula: C9H18O5S Formula Weight: 238.30 Storage Temperature: -30°C to -10°C Melting Point: 109-115° Beilstein Reference: 4631 Optical Rotation: -32° (c=1 in water) MDL No.: MFCD00063273
Formel: C₉H₁₈O₅S
Molecular Weight: 238,31 g/mol
Siedepunkt: 402 °C (1013 hPa)
Schmelzpunkt: 109…115 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00063273
CAS-Nummer: 367-93-1
EINECS: 206-703-0