IPTG (Isopropyl-β-D-thiogalactopyranosid)

Lieferant: Uptima

Synonyme: IPTG

UP84853C UP84853D
UPTIUP84853CEA 0 EUR
UPTIUP84853C UPTIUP84853D
IPTG (Isopropyl-β-D-thiogalactopyranosid)
IPTG (Isopropyl-β-D-thiogalactopyranosid)
Formel: C₉H₁₈O₅S
Molecular Weight: 238.31 g/mol
Siedepunkt: 402
Schmelzpunkt: 109.5…110.5 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00063273
CAS-Nummer: 367-93-1
EINECS: 206-703-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR