IPTG (Isopropyl-β-D-thiogalactopyranosid) Enzymsubstrat dioxanfrei
Lieferant: Thermo Fisher Scientific
Synonyme: IPTG
R0392 R0393
R0392EA 87.75 EUR
R0392 R0393
IPTG (Isopropyl-β-D-thiogalactopyranosid) Enzymsubstrat dioxanfrei
IPTG (Isopropyl-β-D-thiogalactopyranosid)
The IPTG (isopropyl-beta-D-thiogalactopyranoside) is a highly stable synthetic analog of lactose
It inactivates the lac repressor and induces synthesis of beta-galactosidase, an enzyme that promotes lactose utilization. IPTG is used to induce expression of cloned genes under control of the lac operon It is used in conjunction with X-Gal to determine the lac phenotype in blue or white colony screening.
Formel: C₉H₁₈O₅S
Molecular Weight: 238,31 g/mol
Siedepunkt: 402 °C (1013 hPa)
Schmelzpunkt: 109,5…110.5 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00063273
CAS-Nummer: 367-93-1
EINECS: 206-703-0