L-Phenylalaninol 98%

Lieferant: Thermo Fisher Scientific
Gefahr

183870010 183870050 183870250
ACRO183870010EA 30.4 EUR
ACRO183870010 ACRO183870050 ACRO183870250
L-Phenylalaninol 98%
L-Phenylalaninol
Formel: C₉H₁₃NO
Siedepunkt: 278 °C (1013 hPa)
Schmelzpunkt: 90…94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004732
CAS-Nummer: 3182-95-4
EINECS: 221-674-4
UN: 3259
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 90 to 94 °C
Specific optical rotation -21° to -25° (22 °C, 589 nm) (c=1.2, 1 N HCl)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR