Guaifenesin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Guajacolglycerinether, 3-(2-Methoxyphenoxy)-1,2-propandiol, 3-(2-Methoxyphenoxy)propan-1,2-diol, 1-(2,3-Dihydroxypropoxy)-2-methoxybenzol

G5627-500G G5627-25G
SIALG5627-500GEA 300 EUR
SIALG5627-500G SIALG5627-25G
Guaifenesin, Sigma-Aldrich®
Guaifenesin
Formel: C₁₀H₁₄O₄
Molecular Weight: 198,22 g/mol
MDL: MFCD00016873
CAS-Nummer: 93-14-1
EINECS: 202-222-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR