Sudanorange G, rein
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-(Phenylazo)resorcin, Solvent Orange 1
190170050
190170250
190170010
190171000
ACRO190170050EA
23
EUR
ACRO190170050
ACRO190170250
ACRO190170010
ACRO190171000
Sudanorange G, rein
Sudan Orange G
Formel:
C₁₂H₁₀N₂O₂ Molecular Weight: 214,22 g/mol Siedepunkt: 371 °C (1013 hPa) Schmelzpunkt: 143…146 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002275 CAS-Nummer: 2051-85-6 EINECS: 218-131-9 |
Spezifikation Testergebnisse
Appearance | Orange-red Powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...