1-Brom-3-(difluormethoxy)benzol
Lieferant: FLUOROCHEM
007863-5G
007863-10G
FLUO007863-5GEA
66.2
EUR
FLUO007863-5G
FLUO007863-10G
1-Brom-3-(difluormethoxy)benzol
1-Brom-3-(difluormethoxy)benzol
Formel:
C₇H₅BrF₂O Molecular Weight: 223,02 g/mol |
MDL:
MFCD00236233 CAS-Nummer: 262587-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...