1-Brom-3,3,3-trifluor-2-propanol

Lieferant: FLUOROCHEM

Synonyme: 3-Brom-1,1,1-trifluor-2-propanol

001229-1G 001229-5G
FLUO001229-1GEA 63.9 EUR
FLUO001229-1G FLUO001229-5G
1-Brom-3,3,3-trifluor-2-propanol
1-Bromo-3,3,3-trifluoro-2-propanol
Formel: BrH₂CCH(OH)CF₃
Molecular Weight: 192.96 g/mol
MDL: MFCD00040945
CAS-Nummer: 431-34-5
UN: 1987
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR