Chlorphenesin ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

A17386.14 A17386.22 A17386.36
A17386.14EA 45 EUR
A17386.14 A17386.22 A17386.36
Chlorphenesin ≥99%
Chlorphenesin

Purity: 99% CAS No.: 104-29-0 Molecular Formula: C9H11ClO3 Formula Weight: 202.64 Melting Point: 78-82° MDL No.: MFCD00021990

Formel: C₉H₁₁ClO₃
Molecular Weight: 202.64 g/mol
Schmelzpunkt: 78…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00021990
CAS-Nummer: 104-29-0
EINECS: 203-192-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR