3-Acetylbenzonitril ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Methyl-m-cyanphenylketon, 3'-Cyanacetophenon

A10841.06 A10841.14
A10841.06EA 60.8 EUR
A10841.06 A10841.14
3-Acetylbenzonitril ≥97%
3-Acetylbenzonitril

Purity: 97+% CAS No.: 6136-68-1 Molecular Formula: C9H7NO Formula Weight: 145.16 Melting Point: 97-101° Boiling Point: 120°/5mm UN No.: 3439 MDL No.: MFCD00001806

Formel: C₉H₇NO
Molecular Weight: 145.16 g/mol
Siedepunkt: 120 °C (5 mmHg)
Schmelzpunkt: 97…101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001806
CAS-Nummer: 6136-68-1
EINECS: 228-110-6
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR