3-Acetylbenzonitril

Lieferant: FLUOROCHEM

Synonyme: Methyl-m-cyanphenylketon, 3'-Cyanacetophenon

001255-5G 001255-25G
FLUO001255-5GEA 36.9 EUR
FLUO001255-5G FLUO001255-25G
3-Acetylbenzonitril
3-Acetylbenzonitril
Formel: C₉H₇NO
Molecular Weight: 145.16 g/mol
Siedepunkt: 120 °C (5 mmHg)
Schmelzpunkt: 98…100 °C
MDL: MFCD00001806
CAS-Nummer: 6136-68-1
EINECS: 228-110-6
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR