4-Acetylbenzonitril

Lieferant: Thermo Scientific

Synonyme: 4'-Cyanacetophenon, Methyl-p-cyanphenylketon

TL00209.1 TL00209.10 TL00209.50
MAYBTL00209.1EA 0 EUR
MAYBTL00209.1 MAYBTL00209.10 MAYBTL00209.50
4-Acetylbenzonitril
4-Acetylbenzonitril
Formel: C₉H₇NO
Molecular Weight: 145.16
Siedepunkt: 110…112 °C (1 mmHg)
Schmelzpunkt: 57…58 °C
MDL: MFCD00001825
CAS-Nummer: 1443-80-7
EINECS: 215-885-0
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR