1-Ethylpyrrol
Lieferant: FLUOROCHEM
Synonyme:
N-Ethylpyrrole
002566-5G
002566-25G
FLUO002566-5GEA
48.8
EUR
FLUO002566-5G
FLUO002566-25G
1-Ethylpyrrol
1-Ethylpyrrole
Formel:
C₆H₉N Molecular Weight: 95,14 g/mol |
MDL:
MFCD00060633 CAS-Nummer: 617-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...