1-(2-Thenoyl)-3,3,3-trifluoraceton, blassgelbes kristallines Pulver
Lieferant: MP Biomedicals
Achtung
Synonyme: (2-Thenoyl)-ω,ω,ω-trifluoraceton, 2-(4,4,4-Trifluor-1,3-butandionyl)thiophen, 4,4,4-Trifluor-1-(2-thienyl)butan-1,3-dion, TTFA, 2-Thenoyltrifluoraceton
0215684225 0215684280 0215684210
ICNA0215684225EA 85.9 EUR
ICNA0215684225 ICNA0215684280 ICNA0215684210
1-(2-Thenoyl)-3,3,3-trifluoraceton, blassgelbes kristallines Pulver
1-(2-Thenoyl)-3,3,3-trifluoraceton
Thenoyltrifluoroacetone is used as a reagent for the extraction and spectrophotometric determination of metals, actinides and lanthanides. A metal ion chelator that acts as an inhibitor of electron transport complex II in the mitochondrial respiratory chain. Also acts as an inhibitor of succinate and maleate dehydrogenases.
Thenoyltrifluoroacetone is used as a reagent for the extraction and spectrophotometric determination of metals, actinides and lanthanides. A metal ion chelator that acts as an inhibitor of electron transport complex II in the mitochondrial respiratory chain. Also acts as an inhibitor of succinate and maleate dehydrogenases.
Solubility in Methanol (transparency) and 95% ethanol (50 mg/ml - clear to very slightly hazy, colourless to orange solution).
Formel: C₈H₅F₃O₂S
Molecular Weight: 222.19 g/mol
Siedepunkt: 227
Schmelzpunkt: 40…44 °C (Lit.)
Flammpunkt: 112 °C
MDL: MFCD00005445
CAS-Nummer: 326-91-0
EINECS: 206-316-7
Bestellen
Spezifikation Testergebnisse
Identity Test Passes
FTIR Conforms to Standard