Sudan III
Lieferant: Thermo Fisher Scientific
Synonyme:
Sudan Rot, Sudan Red BK, Sudan® III (Rot), 1-(4-(Phenylazo)phenylazo)-2-naphthol, Tony Red, Scarlet B, 1-([4-[(Phenyl)diazenyl]phenyl]diazenyl)naphthalen-2-ol, Fat Ponceau G, Sudan G, Cerasin Red, Sudan Red B, Organol® Rot BS, Sudan Red III, Sudan V
A18318.22
A18318.14
A18318.36
A18318.22EA
56.2
EUR
A18318.22
A18318.14
A18318.36
Sudan III
Sudan III
CAS No.: 85-86-9
Molecular Formula: C22H16N4O
Formula Weight: 352.40
Melting Point: ca 200° dec.
Beilstein Reference: 931185
Merck Reference: 14,8884
MDL No.: MFCD00003905
Formel:
C₂₂H₁₆N₄O Molecular Weight: 352.4 g/mol Schmelzpunkt: ∼ 200 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003905 CAS-Nummer: 85-86-9 EINECS: 201-638-4 Merck Index: 12,09054 |