Pyridoxin Hydrochlorid ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
Adermine hydrochloride, Vitamin B6 hydrochloride, PN HCl, Pyridoxine HCl, (5-Hydroxy-6-methylpyridine-3,4-diyl)dimethanol hydrochloride, Pyridoxol hydrochloride, Pyridoxine HCI
A12041.14
A12041.22
A12041.36
A12041.14EA
39.2
EUR
A12041.14
A12041.22
A12041.36
Pyridoxin Hydrochlorid ≥99%
Pyridoxin Hydrochlorid
Purity: 99%
CAS No.: 58-56-0
Molecular Formula: C8H11NO3.HCl
Formula Weight: 205.64
Odour: Odourless
pH: 3
Melting Point: ca 206° dec.
Beilstein Reference: 3632435
Merck Reference: 14,7982
MDL No.: MFCD00012807
Formel:
C₈H₁₂ClNO₃ Molecular Weight: 205.64 g/mol Schmelzpunkt: ∼ 206 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012807 CAS-Nummer: 58-56-0 EINECS: 200-386-2 Merck Index: 13,08072 |