Pyridoxin Hydrochlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Adermine hydrochloride, Vitamin B6 hydrochloride, PN HCl, Pyridoxine HCl, (5-Hydroxy-6-methylpyridine-3,4-diyl)dimethanol hydrochloride, Pyridoxol hydrochloride, Pyridoxine HCI
M-Clarity™ Qualitätsstufe = MQ200
P9755-25G
P9755-100G
SIALP9755-25GEA
112
EUR
SIALP9755-25G
SIALP9755-100G
Pyridoxin Hydrochlorid, Sigma-Aldrich®
Pyridoxin Hydrochlorid
Formel:
C₈H₁₂ClNO₃ Molecular Weight: 205.64 g/mol Schmelzpunkt: 214…215 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012807 CAS-Nummer: 58-56-0 EINECS: 200-386-2 Merck Index: 13,08072 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...