5-Chlor-2-fluorbenzoylchlorid

Lieferant: FLUOROCHEM

Synonyme: 5-Chlor-2-fluorbenzol-1-carbonylchlorid

CFF945. 005622-5G 005622-10G 005622-25G
CHMPCFF945.EA 0 EUR
CHMPCFF945. FLUO005622-5G FLUO005622-10G FLUO005622-25G
5-Chlor-2-fluorbenzoylchlorid
5-Chlor-2-fluorbenzoylchlorid
Formel: C₇H₃Cl₂FO
Molecular Weight: 193 g/mol
Siedepunkt: 103…104 °C (15 mmHg)
Dichte: 1.459 g/cm³ (20 °C)
MDL: MFCD01631433
CAS-Nummer: 394-29-6
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR