4,5-Dibrom-2-thenoesäure
Lieferant: Thermo Scientific
Synonyme:
4,5-Dibromthiophen-2-carbonsäure, 4,5-Dibrom-2-thiophencarbonsäure
BTB12603.1
BTB12603.10
BTB12603.50
MAYBBTB12603.1EA
0
EUR
MAYBBTB12603.1
MAYBBTB12603.10
MAYBBTB12603.50
4,5-Dibrom-2-thenoesäure
4,5-Dibrom-2-thenoesäure
Formel:
C₅H₂Br₂O₂S Molecular Weight: 285,94 g/mol |
MDL:
MFCD00039682 CAS-Nummer: 6324-10-3 EINECS: 228-683-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...