5-Brom-2-thiophensulfonylchlorid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A14677.14
A14677.06
A14677.14EA
310
EUR
A14677.14
A14677.06
5-Brom-2-thiophensulfonylchlorid ≥98%
5-Brom-2-thiophensulfonylchlorid
Purity: 98%
CAS No.: 55854-46-1
Molecular Formula: C4H2BrClO2S2
Formula Weight: 261.55
Melting Point: 39-42°
Boiling Point: 99-101°/0.5mm
UN No.: 3261
MDL No.: MFCD00084909
Formel:
C₄H₂BrClO₂S₂ Siedepunkt: 99…101 °C (0,5 mmHg) Schmelzpunkt: 39…42 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084909 CAS-Nummer: 55854-46-1 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...