5-Brom-2-thiophensulfonylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Brom-2-thiophensulfonsäurechlorid, 5-Bromthiophen-2-sulfonylchlorid

M-Clarity™ Qualitätsstufe = MQ100

636223-25G 636223-5G
SIAL636223-25GEA 510 EUR
SIAL636223-25G SIAL636223-5G
5-Brom-2-thiophensulfonylchlorid, Sigma-Aldrich®
5-Brom-2-thiophensulfonylchlorid
Formel: C₄H₂BrClO₂S₂
Molecular Weight: 261,55 g/mol
MDL: MFCD00084909
CAS-Nummer: 55854-46-1
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR