5-Brom-2-thiophensulfonylchlorid

Lieferant: CHEMPUR

Synonyme: 5-Brom-2-thiophensulfonsäurechlorid, 5-Bromthiophen-2-sulfonylchlorid

KM03757.250 KM03757DA
CHMPKM03757.250EA 0 EUR
CHMPKM03757.250 MAYBKM03757DA
5-Brom-2-thiophensulfonylchlorid
5-Brom-2-thiophensulfonylchlorid
Formel: C₄H₂BrClO₂S₂
Molecular Weight: 261,55 g/mol
MDL: MFCD00084909
CAS-Nummer: 55854-46-1
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR