Coomassie® Brillantblau R 250, rein, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
Brillantblau R, Hydrogen[4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulfonatobenzyl)amino]benzhydrylen]cyclohexa-2,5-dien-1-yliden](ethyl)(m-sulfonatobenzyl)ammonium Mononatriumsalz, Coomassie Blau R 250, Xylenbrillantcyanin, Serva Blue R, Brillantindocyanin 6B, Acid Blue 83
M-Clarity™ Qualitätsstufe = MQ200
B7920-10G
B7920-50G
B7920-10GEA
118
EUR
B7920-10G
B7920-50G
Coomassie® Brillantblau R 250, rein, Sigma-Aldrich®
Coomassie® Brillantblau R 250
Brilliant Blue R has been used in staining of gels after electrophoresis. It has also been used to determine protein in the protein bodies. For detection of protein bands following electrophoresis.
- Brilliant Blue R is a methanol based stain
- It can also stain agarose gels
- It consists of methanol and acetic acid
Formel:
C₄₅H₄₄N₃NaO₇S₂ Molecular Weight: 825,98 g/mol Schmelzpunkt: 174…180 °C Lagertemperatur: Room Temperature |
MDL:
MFCD00041762 CAS-Nummer: 6104-59-2 EINECS: 228-060-5 |
Spezifikation Testergebnisse
Appearance (Form) | Powder |
Infrared spectrum | Conforms to Structure |
Lambda Max in EtOH |
586 - 592 nm |
Solubility (Colour) 1 mg/ml H₂O |
Dark Blue |
Purity (HPLC) | ≥ 70 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...