3,4-Diethoxy-3-cyclobuten-1,2-dion

Lieferant: Thermo Scientific

Synonyme: Diethylsquarat

RDP00117.1 RDP00117EA
MAYBRDP00117.1EA 0 EUR
MAYBRDP00117.1 MAYBRDP00117EA
3,4-Diethoxy-3-cyclobuten-1,2-dion
3,4-Diethoxy-3-cyclobuten-1,2-dion
Formel: C₈H₁₀O₄
Molecular Weight: 170,16 g/mol
Siedepunkt: 88…92 °C (0,4 mmHg)
Dichte: 1,15 g/cm³ (20 °C)
MDL: MFCD00001333
CAS-Nummer: 5231-87-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR