Diethoxy(diethyl)silan

Lieferant: CHEMPUR SILANE

SILS06375.10 SILS06375.25
SILNSILS06375.10EA 0 EUR
SILNSILS06375.10 SILNSILS06375.25
Diethoxy(diethyl)silan
Diethoxy(diethyl)silan
Formel: C₈H₂₀O₂Si
MDL: MFCD00026755
CAS-Nummer: 5021-93-2
EINECS: 225-706-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR