Diethoxy(dimethyl)silan
Lieferant: CHEMPUR SILANE
Synonyme:
Dimethyldiethoxysilan
SILS07125.25
SILS07125.100
SILNSILS07125.25EA
0
EUR
SILNSILS07125.25
SILNSILS07125.100
Diethoxy(dimethyl)silan
Diethoxy(dimethyl)silan
Formel:
C₆H₁₆O₂Si Molecular Weight: 148.28 g/mol Siedepunkt: 114 Schmelzpunkt: –87 °C Dichte: 0.853 g/cm³ (20 °C) |
MDL:
MFCD00009068 CAS-Nummer: 78-62-6 EINECS: 201-127-6 UN: 2380 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...