Diethoxy(dimethyl)silan ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Dimethyldiethoxysilan
L14725.18
L14725.30
L14725.0B
L14725.18EA
27.8
EUR
L14725.18
L14725.30
L14725.0B
Diethoxy(dimethyl)silan ≥97%
Diethoxy(dimethyl)silan
Purity: 97%
CAS No.: 78-62-6
Molecular Formula: C6H16O2Si
Formula Weight: 148.28
Flash Point: 11°(52°F)
Refractive Index: 1.3820
Odour: Mild
Melting Point: -70°
Boiling Point: 111-113°
Beilstein Reference: 1736110
UN No.: 2380
MDL No.: MFCD00009068
Formel:
C₆H₁₆O₂Si Molecular Weight: 148.28 g/mol Siedepunkt: 111…113 °C Schmelzpunkt: –70 °C Dichte: 0.865 Flammpunkt: 11 °C (52 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009068 CAS-Nummer: 78-62-6 EINECS: 201-127-6 UN: 2380 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...