Diethoxy(diphenyl)silan ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Diphenyldiethoxysilan
B22204.18
B22204.30
B22204.18EA
99.8
EUR
B22204.18
B22204.30
Diethoxy(diphenyl)silan ≥98%
Diethoxy(diphenyl)silan
Purity: 98%
CAS No.: 2553-19-7
Molecular Formula: C16H20O2Si
Formula Weight: 272.42
Flash Point: 175°(347°F)
Refractive Index: 1.5255
Boiling Point: 166-168°/15mm
Beilstein Reference: 2281302
MDL No.: MFCD00015126
Formel:
C₁₆H₂₀O₂Si Molecular Weight: 272,42 g/mol Siedepunkt: 166…168 °C (15 mmHg) Dichte: 1,033 Flammpunkt: 175 °C (347 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015126 CAS-Nummer: 2553-19-7 EINECS: 219-860-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...