Diethoxy(diphenyl)silan

Lieferant: CHEMPUR SILANE

SILS07825.25 SILS07825.100
SILNSILS07825.25EA 0 EUR
SILNSILS07825.25 SILNSILS07825.100
Diethoxy(diphenyl)silan
Diethoxy(diphenyl)silan
Formel: C₁₆H₂₀O₂Si
MDL: MFCD00015126
CAS-Nummer: 2553-19-7
EINECS: 219-860-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR