Dimethoxy(dimethyl)silan ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Dimethyldimethoxysilan

A12025.14 A12025.22
A12025.14EA 47.6 EUR
A12025.14 A12025.22
Dimethoxy(dimethyl)silan ≥97%
Dimethoxy(dimethyl)silan

Purity: 97% CAS No.: 1112-39-6 Molecular Formula: C4H12O2Si Formula Weight: 120.22 Flash Point: -8°(17°F) Refractive Index: 1.3710 Melting Point: -80° Boiling Point: 81-82° Beilstein Reference: 1733694 UN No.: 1993 MDL No.: MFCD00025691

Formel: C₄H₁₂O₂Si
Molecular Weight: 120,22 g/mol
Siedepunkt: 81…82 °C
Schmelzpunkt: –80 °C
Dichte: 0,867
Flammpunkt: –8 °C (17 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00025691
CAS-Nummer: 1112-39-6
EINECS: 214-189-4
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR