Dimethoxy(dimethyl)silan
Lieferant: CHEMPUR SILANE
Synonyme:
Dimethyldimethoxysilan
SILS07200.25
SILS07200.100
SILS07200.250
SILNSILS07200.25EA
0
EUR
SILNSILS07200.25
SILNSILS07200.100
SILNSILS07200.250
Dimethoxy(dimethyl)silan
Dimethoxy(dimethyl)silan
Formel:
C₄H₁₂O₂Si Molecular Weight: 120,22 g/mol Siedepunkt: 82 °C (1013 hPa) Schmelzpunkt: –80 °C Dichte: 0,8638 g/cm³ (20 °C) |
MDL:
MFCD00025691 CAS-Nummer: 1112-39-6 EINECS: 214-189-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...