1-Iod-7-(trifluormethyl)perfluoroctan

Lieferant: FLUOROCHEM

007077-25G 007077-100G
FLUO007077-25GEA 0 EUR
FLUO007077-25G FLUO007077-100G
1-Iod-7-(trifluormethyl)perfluoroctan
1-Iod-7-(trifluormethyl)perfluoroctan
Formel: C₉F₁₉I
Molecular Weight: 595.97 g/mol
MDL: MFCD00153243
CAS-Nummer: 865-77-0
EINECS: 212-747-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR