Montage® Plasmid Miniprep 96 Kits
Lieferant: Merck
LSKP09604 LSKP09624
MILFLSKP09604EA 0 EUR
MILFLSKP09604 MILFLSKP09624
Montage® Plasmid Miniprep 96 Kits
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
Plasmid Miniprep 96 kit is a quick and easy-to-use kit that produces clean and reproducible DNA in half a time than traditional methods.
  • No centrifugation or precipitation steps required
  • Minimum processing time
  • Excellent purity, yields, and reproducibility
  • Automation compatible
Bind-wash-elute or centrifugation steps are not required. The kit is inclusive of all the reagents and disposable materials needed to purify plasmid DNA in a 96-well format. Purified samples are applicable for use in most of the sensitive downstream applications such as cloning, DNA sequencing, and transformation.
Application:
Plasmid Miniprep 96 kit has been used to prepare DNA of the expression plasmids for automated apoptosis assay to detect activation of caspase 3.
Lieferumfang: Components includes 96-well filter plates and all the reagents and disposable materials you need to purify plasmid or BAC DNA.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable
Anwendung
Target molecule
Refine target molecule
Sample type
Method / Format