2-Chlor-4-(trifluormethyl)benzonitril

Lieferant: Thermo Scientific

Synonyme: 2-Chlor-4-(trifluormethyl)benzoesäurenitril, 3-Chlor-4-cyanbenzotrifluorid

SEW01274.250 SEW01274.1
MAYBSEW01274.250EA 0 EUR
MAYBSEW01274.250 MAYBSEW01274.1
2-Chlor-4-(trifluormethyl)benzonitril
2-Chlor-4-(trifluormethyl)benzonitril
Formel: C₈H₃ClF₃N
Molecular Weight: 205.57
MDL: MFCD00084939
CAS-Nummer: 1813-33-8
UN: 3276
ADR: 6.1 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR