3,4'-Dichlorpropiophenon ≥97%

Lieferant: Thermo Scientific

A13311.09 A13311.18
A13311.09EA 55.6 EUR
A13311.09 A13311.18
3,4'-Dichlorpropiophenon ≥97%
3,4'-Dichlorpropiophenon

Purity: 97% CAS No.: 3946-29-0 Molecular Formula: C9H8Cl2O Formula Weight: 203.07 Flash Point: 110°(230°F) Melting Point: 48-52° Boiling Point: 135-137°/0.6mm Beilstein Reference: 1866915 MDL No.: MFCD00000992

Formel: C₉H₈Cl₂O
Molecular Weight: 203,07 g/mol
Siedepunkt: 135…137 °C (0,6 mmHg)
Schmelzpunkt: 51 °C
Flammpunkt: >110 °C (230 °F)
MDL: MFCD00000992
CAS-Nummer: 3946-29-0
EINECS: 223-537-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR